ContactUsLinkComponent
Model technology: Filtration ductless fume hood Captair 392 Smart Filters configuration: 1C Detection: A Back panel: Standard Fron t and Side panel: Sequential opening + left waste port Power supply: GB Version: 1 1 * 1 items
Catalog # ERLAAA636A02013A31
Supplier:  ERLAB CAPTAIRE
New Product
Model technology: Filtration ductless fume hood Captair 392 Smart Filters configuration: 1C Detection: A Back panel: Standard Fron t and Side panel: Sequential opening + left waste port Power supply: GB Version: 1 1 * 1 items
Catalog # ERLAAA636A02013A31
Supplier:  ERLAB CAPTAIRE