ContactUsLinkComponent
Polyethylene Glycol 2000 Monomethyl Ether, commonly referred to as PEG 2000 MONOMETHYLETHER, is a polyether compound that is used in a wide variety of fields including pharmaceutical manufacturing as an excipient and active ingredient. Due to its low 1 * 1 Each
Catalog # SPCMP2070-1KG
Supplier:  Spectrum Chemical
New Product
Polyethylene Glycol 2000 Monomethyl Ether, commonly referred to as PEG 2000 MONOMETHYLETHER, is a polyether compound that is used in a wide variety of fields including pharmaceutical manufacturing as an excipient and active ingredient. Due to its low 1 * 1 Each
Catalog # SPCMP2070-1KG
Supplier:  Spectrum Chemical