ContactUsLinkComponent
Muffle furnace l5/13/b510 tmax1300 with flap door protective gas connection and thermocouple hole in the furnace door 1 * 1 items
Catalog # NABEL-055K3ANLGY
Supplier:  NABERTHERM
New Product
Muffle furnace l5/13/b510 tmax1300 with flap door protective gas connection and thermocouple hole in the furnace door 1 * 1 items
Catalog # NABEL-055K3ANLGY
Supplier:  NABERTHERM