Ag, Al, B, Ba, Bi, Ca, Cd, Cr, Cu, Fe, In, Li, K, Mg, Mn, Mo, Na, Ni, P, Pb, Sb, Si, Sn, Ti, V, Zn @ 100 µg/g in 75 cSt Hydrocarbon Oil 1 * 100 g
Catalog # PROOGV26-100-100G
Supplier: LGC Standards PROMOCHEM
New Product
Ag, Al, B, Ba, Bi, Ca, Cd, Cr, Cu, Fe, In, Li, K, Mg, Mn, Mo, Na, Ni, P, Pb, Sb, Si, Sn, Ti, V, Zn @ 100 µg/g in 75 cSt Hydrocarbon Oil 1 * 100 g
Catalog # PROOGV26-100-100G
Supplier: LGC Standards PROMOCHEM
New Product
Frequently Bought Together
Potassium dichromate, Certipur®, Supelco®
Supplier: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Ammonia solution 17% Reag. Ph. Eur. 1004701
Supplier: VWR Chemicals
pH buffer solutions, NIST traceable, Orion™
Supplier: Thermo Orion
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR® Buffer Solutions, Secondary Standards
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
VWR® ARISTAR®, Sodium 1000 mg/L in Water Standard for Ion Chromatography
Supplier: VWR Chemicals
Conductivity standards, KCl, referenced at 25 °C, for conductivity measurement
Supplier: Mettler - Toledo
VWR®, Turbidity standards, formazin
Supplier: VWR Chemicals