REPROSIL 100 C8 3 M / 50 X 3 MM 1 * 1 items
Catalog # MAISR13.86.S0503
Supplier: DR MAISCH
New Product
REPROSIL 100 C8 3 M / 50 X 3 MM 1 * 1 items
Catalog # MAISR13.86.S0503
Supplier: DR MAISCH
New Product
Frequently Bought Together
Safety canisters
Supplier: S.C.A.T.
VWR®, Septa GL 45
Supplier: VWR Collection
VWR®, Screw Caps for Screw Neck Vials ND8
Supplier: VWR Collection
VWR®, Safety Waste Caps
Supplier: VWR Collection
Gastight® 1700 series syringes
Supplier: HAMILTON BONADUZ
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Micro Syringes Ultra-minute Volume
Supplier: VWR Collection
VWR® Gas-Tight Micro Syringes
Supplier: VWR Collection
VWR®, Storage Boxes for Sample Vials and Bottles, PP
Supplier: VWR Collection
Glass screw top vials, 2 ml, level 2 high-throughput applications, SureSTART™
Supplier: Thermo Fisher Scientific
Gas-tight syringes, 10 to 500 µl
Supplier: Trajan Scientific and Medical
VWR® Septa ND8
Supplier: VWR Collection
WHEATON® Aluminium Seals, Unlined
Supplier: DWK Life Sciences
Tubing for peristaltic pumps
Supplier: PerkinElmer