This Sorbitol Dehydrogenase Assay Kit is a quantitative assay kit for measuring Sorbitol Dehydrogenase activity in animal tissue, pl ant tissue, cells, bacteria, serum, and plasma. 1 * 96 Tests
Catalog # ANTIA319736-96
Supplier: ANTIBODIES.COM