ContactUsLinkComponent
This Proline Assay Kit is a quantitative assay kit for measuring Proline in plant tissues, animal tissues, cells, bacteria, serum, a nd plasma. 1 * 48 Tests
Catalog # ANTIA319722-48
Supplier:  ANTIBODIES.COM
New Product
This Proline Assay Kit is a quantitative assay kit for measuring Proline in plant tissues, animal tissues, cells, bacteria, serum, a nd plasma. 1 * 48 Tests
Catalog # ANTIA319722-48
Supplier:  ANTIBODIES.COM