This Monoamine Oxidase Activity Assay Kit is a quantitative assay kit for measuring Monoamine Oxidase / MAO activity in serum, plasm a, animal and plant tissues, cells, and bacteria. 1 * 96 Tests
Catalog # ANTIA319706-96
Supplier: ANTIBODIES.COM