This Lactate Dehydrogenase Assay Kit is a quantitative assay kit for measuring Lactate Dehydrogenase activity in animal and plant ti ssues, cells, plasma, serum, and other liquid samples. 1 * 480 Tests
Catalog # ANTIA319695-480
Supplier: ANTIBODIES.COM