ContactUsLinkComponent
This Glutathione Peroxidase Activity Assay Kit is a quantitative assay kit for measuring Glutathione Peroxidase activity in serum, p lasma, plant tissue/cell lysates, and other biological fluids. 1 * 48 Tests
Catalog # ANTIA319686-48
Supplier:  ANTIBODIES.COM
New Product
This Glutathione Peroxidase Activity Assay Kit is a quantitative assay kit for measuring Glutathione Peroxidase activity in serum, p lasma, plant tissue/cell lysates, and other biological fluids. 1 * 48 Tests
Catalog # ANTIA319686-48
Supplier:  ANTIBODIES.COM