ContactUsLinkComponent
This Oxidized Glutathione (GSSG) Assay Kit is a quantitative assay kit for measuring Oxidized Glutathione in serum, plasma, animal/p lant tissues, blood cells, cells, and bacteria. 1 * 96 Tests
Catalog # ANTIA319685-96
Supplier:  ANTIBODIES.COM
New Product
This Oxidized Glutathione (GSSG) Assay Kit is a quantitative assay kit for measuring Oxidized Glutathione in serum, plasma, animal/p lant tissues, blood cells, cells, and bacteria. 1 * 96 Tests
Catalog # ANTIA319685-96
Supplier:  ANTIBODIES.COM