Novel Juice PLUS is a fluorescent reagent that produces instant visualization of DNA bands upon blue light or UV illumination of aga rose gels. Supplied in 6X DNA Loading Buffer, Novel Juice PLUS is used to prepare DNA markers or samples for loading 1 * 1 mL
Catalog # ANTIA318985-1
Supplier: Antibodies.com