ContactUsLinkComponent
Pre-stained protein ladder. Covers the size range 10-245 kDa (Tris-glycine-SDS running buffer). Number of bands = 12. Green band at approximately 25 kDa. Red band at approximately 75 kDa. Designed for monitoring SDS-PAGE separation and WB transfer ef 1 * 500 µl
  ANTIA318979-500
New Product
Pre-stained protein ladder. Covers the size range 10-245 kDa (Tris-glycine-SDS running buffer). Number of bands = 12. Green band at approximately 25 kDa. Red band at approximately 75 kDa. Designed for monitoring SDS-PAGE separation and WB transfer ef 1 * 500 µl
  ANTIA318979-500