ContactUsLinkComponent
OneMark 100 with Novel Green is designed to show virtually uniform spacing over a wide fragment range. The ladder is supplied in a r eady-to-use format containing the fluorescent DNA stain and tracking dyes. Optimized for direct loading onto unstaine 1 * 600 µl
Catalog # ANTIA318975-600
Supplier:  ANTIBODIES.COM
New Product
OneMark 100 with Novel Green is designed to show virtually uniform spacing over a wide fragment range. The ladder is supplied in a r eady-to-use format containing the fluorescent DNA stain and tracking dyes. Optimized for direct loading onto unstaine 1 * 600 µl
Catalog # ANTIA318975-600
Supplier:  ANTIBODIES.COM