ContactUsLinkComponent
VICIC75NT-1673EUHB - 3 mpos trapping-path - u-electric/USB - 1/16*.25mm, 50C/10000psi liq, N60C/E3 - Electric Universal Actuator - serial interface: USB 1 * 1 items
Catalog # VICIC75NT-1673EUHB
Supplier:  VICI
New Product
VICIC75NT-1673EUHB - 3 mpos trapping-path - u-electric/USB - 1/16*.25mm, 50C/10000psi liq, N60C/E3 - Electric Universal Actuator - serial interface: USB 1 * 1 items
Catalog # VICIC75NT-1673EUHB
Supplier:  VICI