ContactUsLinkComponent
LN2 Capacity (L) 4.6 Static Holding Time (days) - 11 Normal Evaporation Rate (liters/day) - 0.4 Liquid Neck Diameter (mm) - 36 Overall Height / Diameter (mm) - 480 x 264 Weight Empty / Full (kg) - 2.8 / 6.5 1 * 1 items
Catalog # TAYW10-152-01-0095
Supplier:  TAYLOR WHARTON
New Product
LN2 Capacity (L) 4.6 Static Holding Time (days) - 11 Normal Evaporation Rate (liters/day) - 0.4 Liquid Neck Diameter (mm) - 36 Overall Height / Diameter (mm) - 480 x 264 Weight Empty / Full (kg) - 2.8 / 6.5 1 * 1 items
Catalog # TAYW10-152-01-0095
Supplier:  TAYLOR WHARTON