Isolator Gloves - Material : CSM - Shape : E10 - Size : 7 - Thickness (mm) : 4/10 - Length (mm) : 330 - Bead diameter (mm) : 4 mm - Hand : Ambidextrous 1 * 30 items
PIECE10330Y7E44
: PIERCAN
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.