PRIME-XV Human Fibronectin can be used as a coating cell culture substrate to support optimal attachment and growthof human primary cells, including stem/ progenitor cells. The recommended concentration for this application is typically 1- 5 g/ mL. 1 * 1 L
Catalog # SHBT31002
Supplier: Shenandoah Biotechnology