PRIME-XV T Cell XSFM is a xeno-free and serum-free culture medium that supports the expansion and reactivation of human T cells, as well as the transition from preclinical to clinical applications for immunotherapy therapeutics. Designed and manuf l 1 * 1 L
Catalog # SHBT91141
Supplier: Shenandoah Biotechnology