,,10 components: Ca 600mg/l Na 500mg/l P 600mg/l Mg 200mg/l K 800mg/l Fe 10mg/l Cu 1.25mg/l Zn 6mg/l Mn 1.2mg/l Se 0.3mg/l in HNO3 10% ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 8 months.,, ,,Ambient Temp.,, 1 * 250 mL
Catalog # CMPLRM071604L25
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together