Adenosin-5'-triphosphate disodium salt (ATP) 1 * 10 g
Catalog # ROTHHN35.2
Supplier: Roth Carl
Frequently Bought Together
2-Propanol ≥99.0%, GPR RECTAPUR®
Supplier: VWR Chemicals
n-Hexane, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Acetone ≥99.9%, PESTINORM® Supra Trace for organic trace analysis
Supplier: VWR Chemicals
Tetrahydrofuran, anhydrous 99.9% stabilised, extra pure
Supplier: Thermo Fisher Scientific
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
ETHYLENE GLYCOL ≥98 TECHNICAL 1 * 5 L
Supplier: Roth Carl
FAM Benzine DIN 51635, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
N,N-Dimethylformamide, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
1-Octanol ≥99%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Methanol, extra dry 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Cyclohexane for pesticide residue analysis
Supplier: Thermo Fisher Scientific
Cyclohexane 99.8% for HPLC
Supplier: Thermo Fisher Scientific