Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
 

Nuclear extract from HeLa cells

Nuclear extractor, 100 µl
Catalog # ENZOBMLKI1400100
Supplier:   Enzo Life Sciences
Nuclear extract from HeLa cells
Catalog # ENZOBMLKI1400100
Supplier:  Enzo Life Sciences
CAS Number:  
Nuclear extract from HeLa cells

Nuclear extractor, 100 µl

CAS Number:  
Price
£171.00
Log in for availability
1 * 100 µl
Product Details & Documents

Prepared by high salt extraction of HeLa nuclei (human cervical cancer cell line), this extract is a rich source of HDAC activity, useful for inhibitor screening or immunoprecipitation of HDAC containing complexes. Typically, 0,5 µl or less per well is sufficient for assay with the HDAC FLUOR DE LYS® fluorometric activity assay.

Specifications

Specifications

  • Pk:
    100 µl
  • Description:
    Nuclear extractor
  • Cat. No.:
    ENZOBMLKI1400100
  • Storage buffer:
    Liquid. In 0,1 M KCl, 20 mM HEPES/NaOH, pH 7,9, 20% (v/v) glycerol, 0,2 mM EDTA, 0,5 mM DTT, 0,5 mM PMSF.

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.