ContactUsLinkComponent
Thermometer, based on ASTM type S12C, rod shape, capillary: white coated round, measuring range: -20+102C in 0.2C, blue filling, immersion depth: total, total length: 435 mm, including DAkkS calibration certificate with 2 test points 1 * 1 items
  SCNL72636/04/2P
Thermometer, based on ASTM type S12C, rod shape, capillary: white coated round, measuring range: -20+102C in 0.2C, blue filling, immersion depth: total, total length: 435 mm, including DAkkS calibration certificate with 2 test points 1 * 1 items
  SCNL72636/04/2P
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.