Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Memosens CPS61E CPS61E-18R0/0 (CPS61E-AA6MTU2+JE) 1 * 1 items
Catalog # ENDRCPS61E-18R0/0
Supplier:
Endress+Hauser
Expand image
Memosens CPS61E CPS61E-18R0/0 (CPS61E-AA6MTU2+JE) 1 * 1 items
Catalog # ENDRCPS61E-18R0/0
Supplier:
Endress+Hauser
Frequently Bought Together
VWR® pHenomenal® pH Electrodes
Supplier: VWR Collection
Facial tissues, KLEENEX®
Supplier: KIMBERLY CLARK
VWR® Facial Tissues
Supplier: VWR Collection
Wipes for delicate tasks, Kimtech™ Science
Supplier: KIMBERLY CLARK
Hand towels, KLEENEX®
Supplier: KIMBERLY CLARK
VWR®, Bench Protectors
Supplier: VWR Collection
VWR®, Glove Box Holders
Supplier: VWR Collection
VWR® Spec-Wipe® 3, Cleanroom Wipes, Non Woven
Supplier: VWR Collection
pH electrodes, Sentix® IDS
Supplier: WTW
pH electrodes, SenTix® PLUS
Supplier: WTW
Kitchen rolls, Tork Premium
Supplier: Essity
VWR®, Wipes, Pre-Saturated
Supplier: VWR Collection
VWR® Spec-Wipe® 3, Cleanroom Wipes, Pre-Saturated
Supplier: VWR Collection