NALIDIXIC ACID 98 % FOR BIOCHEMISTRY 1 * 10 g
Catalog # ROTHCN32.1
Supplier: Roth Carl
Frequently Bought Together
Dimethyl sulfoxide EMPLURA 1 * 185 L
Supplier: Merck
Xylene (mixture of isomers) ≥98%, liquid
Supplier: PanReac AppliChem
Piperidine ≥99.5% (GC, on anhydrous basis) for peptide synthesis
Supplier: VWR Chemicals
1,2-Dichlorobenzene 99+% for HPLC
Supplier: Thermo Fisher Scientific
ETHYL ACETATE 99.5% FOR SYNTHESIS 1 * 200 L
Supplier: PanReac AppliChem
2,2,4-Trimethylpentane 99.5% for analysis
Supplier: Thermo Fisher Scientific
Acetonitrile 99.9%, ECD tested for residue analysis
Supplier: Thermo Fisher Scientific
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
n-Hexane 95%, ChromAR®, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Dimethyl Sulfoxide (USP, BP, Ph. Eur.) pharma grade 1 * 190 L
Supplier: PanReac AppliChem
2-Propanol(USP, BP, Ph. Eur.)pure, pharma grade 1 * 200 L
Supplier: PanReac AppliChem