,, ,,1 component: Chlorothalonil metabolite R417888 sodium [CAS:N/A] 100mg/l in Acetonitrile ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 12 months.,, ,,Store under normal laboratory conditions, at temperatures between 15C to 25C,, 1 * 1 mL
Catalog # CMPLRD0923401
Supplier: CUSTOM MADE CHEMICALS LAB