,,1 component: Chloridazon-methyl-desphenyl [CAS:17254-80-7] 100mg/l in Acetonitrile ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 12 months.,, ,,Store under normal laboratory conditions, at temperatures between 15C to 25C,, 1 * 1 mL
Catalog # CMPLRD0782191
Supplier: CUSTOM MADE CHEMICALS LAB