,,1 component: Chloridazon-desphenyl [CAS:6339-19-1] 100mg/l in Acetonitrile ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 12 months.,, ,,Store in a refrigerator at temperatures between 2C to 8C,, 1 * 1 mL
Catalog # CMPLRD0379061
Supplier: CUSTOM MADE CHEMICALS LAB