N-Butyl-2-pyrrolidone (NBP), 100 ml 1 * 100 mL
Catalog # ROTH1E8A.1
Supplier: Roth Carl
Frequently Bought Together
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
m-Xylene for synthesis, Sigma-Aldrich®
Supplier: Merck
Cyclohexanone ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Iodomethane ≥99.5% stabilised
Supplier: Thermo Fisher Scientific
Toluene 99.5% ACS for spectroscopy
Supplier: Thermo Fisher Scientific
tert-Butanol 99.5% for analysis
Supplier: Thermo Fisher Scientific
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Chloroform 99.0-99.4% (by GC) stabilised, Puriss. p.a. ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
1-Propanol ACS for spectroscopy
Supplier: Thermo Fisher Scientific
n-Hexane 95%, ChromAR®, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Isoamyl Alcohol according to Gerber for analysis 1 * 1.000 mL
Supplier: PanReac AppliChem