ContactUsLinkComponent
4 components: 1000mg/l each of In Tb Sc Y in HNO3 5% CRM, ISO 17034 and ISO 17025 Conservation 24 months. (12 months after opening) Store under normal laboratory conditions, at temperatures between 15C to 25C 1 * 125 mL
Catalog # CMPLRM040783L125
4 components: 1000mg/l each of In Tb Sc Y in HNO3 5% CRM, ISO 17034 and ISO 17025 Conservation 24 months. (12 months after opening) Store under normal laboratory conditions, at temperatures between 15C to 25C 1 * 125 mL
Catalog # CMPLRM040783L125
Frequently Bought Together