BOTTLE W/N AMBER 100ML GL40 1 * 63 items
Catalog # 215-4698
Supplier: VWR Collection
Frequently Bought Together
VWR®, Nitrile Gloves
Supplier: VWR Collection
Pipette tips, epT.I.P.S.® Standard (General Lab Product)
Supplier: EPPENDORF
VWR® Facial Tissues
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
BOTTLE W/N AMBER 30ML GL32 1 * 126 items
Supplier: VWR Collection
Hydrogen peroxide 31% VLSI, Electronic Grade
Supplier: VWR Chemicals
Quinoline yellow sulphonic acid sodium salt (Acid yellow 3) 95%, water soluble, pure
Supplier: Thermo Fisher Scientific