BOTTLE W/N AMBER 50ML GL32 1 * 85 items
Catalog # 215-4695
Supplier: VWR Collection
Frequently Bought Together
Pipette tips, Finntip™
Supplier: Thermo Fisher Scientific
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Assistent Brushes with Cotton Tip Bristles
Supplier: GLASWARENFABRIK KARL HECHT
Single-use syringes, 2-piece, HENKE-JECT®
Supplier: HENKE SASS WOLF
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Nitrile gloves, Kimtech™ Blue Nitrile™
Supplier: KIMBERLY CLARK
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
Nitric acid 0.2 mol/l (0.2 N)
Supplier: VWR Collection