ContactUsLinkComponent
28S rRNA (+) Control CISH Probe is designed to be used for the qualitative detection of human 28S ribosomal RNA in formalin-fixed, p araffin-embedded tissue or cells by chromogenic in situ hybridization (CISH). 1 * 100 µl
Catalog # ABNOCO0005.100UL
Supplier:  Abnova
28S rRNA (+) Control CISH Probe is designed to be used for the qualitative detection of human 28S ribosomal RNA in formalin-fixed, p araffin-embedded tissue or cells by chromogenic in situ hybridization (CISH). 1 * 100 µl
Catalog # ABNOCO0005.100UL
Supplier:  Abnova