ContactUsLinkComponent
PEG-BSA Coated 96-Well Plate is coated with purified mono-PEGylated BSA (20 kDa mPEG). It is stabilized with a proprietary blocking/ stabilizer buffer. 1 * 1 items
Catalog # ABNOU0520.1PLATE
Supplier:  Abnova
PEG-BSA Coated 96-Well Plate is coated with purified mono-PEGylated BSA (20 kDa mPEG). It is stabilized with a proprietary blocking/ stabilizer buffer. 1 * 1 items
Catalog # ABNOU0520.1PLATE
Supplier:  Abnova