ContactUsLinkComponent
GTP is Guanosine-5'-triphosphate, which is an extremely stable nucleotide.This compound and its sodium salt are essential components of energy conversion in biological systems and can be used in various related applications in molecular biology, suc 1 * 100 mL
Catalog # ABNOU0509.100ML
Supplier:  Abnova
GTP is Guanosine-5'-triphosphate, which is an extremely stable nucleotide.This compound and its sodium salt are essential components of energy conversion in biological systems and can be used in various related applications in molecular biology, suc 1 * 100 mL
Catalog # ABNOU0509.100ML
Supplier:  Abnova