ContactUsLinkComponent
CTP Tris solution is supplied as a 100 mM/L clear colorless to yellow solution solution. 1 * 100 mL
Catalog # ABNOU0468.100ML
Supplier:  Abnova
CTP Tris solution is supplied as a 100 mM/L clear colorless to yellow solution solution. 1 * 100 mL
Catalog # ABNOU0468.100ML
Supplier:  Abnova