ContactUsLinkComponent
S-Adenosyl Methionine (SAM) is an auxiliary substrate involved in methyl transfer reaction. It is synthesized in vivo by adenosine t riphosphate and methionine under the action of Methionine adenosyltransferase. 1 * 20 mL
Catalog # ABNOU0443.20ML
Supplier:  Abnova
S-Adenosyl Methionine (SAM) is an auxiliary substrate involved in methyl transfer reaction. It is synthesized in vivo by adenosine t riphosphate and methionine under the action of Methionine adenosyltransferase. 1 * 20 mL
Catalog # ABNOU0443.20ML
Supplier:  Abnova