ContactUsLinkComponent
Microscope cover glasses, 13 mm Ø, borosilicate glass D 263® T eco, thickness No. 3 (0.28-0.32 mm) 1 * 500 items
  MARI0114530
Microscope cover glasses, 13 mm Ø, borosilicate glass D 263® T eco, thickness No. 3 (0.28-0.32 mm) 1 * 500 items
  MARI0114530