Beaker 1000 ml, tall shape, with handle and spout, Ø 95 mm, height: 180 mm, with graduation, made from borosilicate glass 3.3 Pack contains 2 pcs/p 1 * 1 Each
WITG5.503.001
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.