SHIELD SAFETY VALVE 110 x 40MM 1 * 1 items
Catalog # TUTTCOV411-0010
Supplier: TUTTNAUER
Frequently Bought Together
Filter protection 5 ml 1 * 50 items
Supplier: EPPENDORF
Accessories for pipetus® / pipetus®-standard / pipetus®-junior
Supplier: HIRSCHMANN
VWR®, Accessories for Mechanical Pipettes
Supplier: VWR Collection
Cover gasket 1 * 1 items
Supplier: VWR Collection
Accessories for HandyStep® electronic
Supplier: Brand
Filter,membrane for research pipette 1 * 50 items
Supplier: EPPENDORF
Accessories for accu-jet® pro
Supplier: Brand
Sacrifical body 1 * 1 items
Supplier: VWR Collection
Accessories for Transferpette® electronic pipettes
Supplier: Brand
Accessories for electronic dispensers, Multipette E3/E3x
Supplier: EPPENDORF
Vacuum seal PTFE / FKM
Supplier: Heidolph Instruments GmbH & Co.KG
BIOTECTOR BASE REAGENT 1 * 20 L
Supplier: Hach
Rotors and accessories for Megafuge® 8 centrifuges
Supplier: Thermo Scientific
ORP combination electrodes, IntelliCAL™ MTC101 / MTC301
Supplier: Hach
Accessories
Supplier: Brand
Accessories for PIPETBOY acu / pro
Supplier: Integra
Accessories for CO2 Incubators, Heracell VIOS
Supplier: Thermo Scientific
SILICONE OIL 1 * 1 items
Supplier: Brand