ContactUsLinkComponent
LONG TIME FREEZER 21 Litres w/6 Canisters Static Holding Time (days) - 227 Normal Evaporation Rate (liters/day) - 0.095 Liquid Nitrogen Capacity - 21.6 litres Neck Diameter (mm) - 55.6 Overall Height / Diameter (mm) - 658 x 368 Weight Empty / Full (k 1 * 1 items
Catalog # TAYW10-152-01-0032
Supplier:  TAYLOR WHARTON
LONG TIME FREEZER 21 Litres w/6 Canisters Static Holding Time (days) - 227 Normal Evaporation Rate (liters/day) - 0.095 Liquid Nitrogen Capacity - 21.6 litres Neck Diameter (mm) - 55.6 Overall Height / Diameter (mm) - 658 x 368 Weight Empty / Full (k 1 * 1 items
Catalog # TAYW10-152-01-0032
Supplier:  TAYLOR WHARTON