ContactUsLinkComponent
Densimeter acc. to Candlot-Le Chatelier, 250 ml, borosilicate glass 3.3, graduated 0 to 1 ml/0,1 ml and marks at 18 to 25 ml/0,1 ml, tolerance 0,05 ml, with funnel and hollow stopper 1 * 1 items
Catalog # WITG3.688.000
Supplier:  witeg Labortechnik
Densimeter acc. to Candlot-Le Chatelier, 250 ml, borosilicate glass 3.3, graduated 0 to 1 ml/0,1 ml and marks at 18 to 25 ml/0,1 ml, tolerance 0,05 ml, with funnel and hollow stopper 1 * 1 items
Catalog # WITG3.688.000
Supplier:  witeg Labortechnik
Frequently Bought Together