ContactUsLinkComponent
Furnace MuFFle lt9/11/skM/b510 tMax1100 with with liFt Door Door anD over-teMPerature liMiter anD therMocouPle hole in the rear wall 1 * 1 items
Catalog # NABELM095H1ANLTZ
Supplier:  NABERTHERM
Furnace MuFFle lt9/11/skM/b510 tMax1100 with with liFt Door Door anD over-teMPerature liMiter anD therMocouPle hole in the rear wall 1 * 1 items
Catalog # NABELM095H1ANLTZ
Supplier:  NABERTHERM
Frequently Bought Together