ContactUsLinkComponent
Furnace MuFFle l5/13/c550 tMax1300 with FlaP Door anD Protective gas connection, over-teMPerature liMiter anD therMocouPle hole in t he rear wall 1 * 1 items
Catalog # NABEL-055K3ANOGTZ
Supplier:  NABERTHERM
Furnace MuFFle l5/13/c550 tMax1300 with FlaP Door anD Protective gas connection, over-teMPerature liMiter anD therMocouPle hole in t he rear wall 1 * 1 items
Catalog # NABEL-055K3ANOGTZ
Supplier:  NABERTHERM
Frequently Bought Together