Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 32 g. Draft Shield D 1 * 1 items
Catalog # SARBMCA36S3CF24
Supplier: Sartorius Balances
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 32 g. Draft Shield D 1 * 1 items
Catalog # SARBMCA36S3CF24
Supplier: Sartorius Balances
Frequently Bought Together
Accessories for dispersing elements for homogenisers
Supplier: IKA
Overhead stirrer tools, propeller, diamond, centrifugal and anchor-type impellers
Supplier: witeg Labortechnik
MS 3 control Orbital schaker 1 * 1 items
Supplier: IKA
Accessories for reciprocating shakers, HS 260
Supplier: IKA
Funnel small ZyklonZM200 1 * 1 items
Supplier: Retsch GmbH
Accessories for variable-speed rotor mill, PULVERISETTE 14
Supplier: FRITSCH
Stirrer shafts, propeller type, BOLA
Supplier: Bohlender
BLENDER STAINLESS CONTAINER 2L MX SERIES 1 * 1 items
Supplier: WARING BLENDOR
HOTPLATE STIRRER MSH-30D-UNIT 260X260M 1 * 1 items
Supplier: witeg Labortechnik
Accessories for KL, KM, KS, SM and VKS shakers
Supplier: Edmund Bühler GmbH
Protective grating with dust filter for fan ZM 300 1 * 1 items
Supplier: Retsch GmbH
VWR®, Multi Purpose Clamps, 3-Prong
Supplier: VWR Collection
CONDENSYN MINI: 200MM L WITH B10 FITTING 1 * 1 items
Supplier: ASYNT
Accessories for boekel tube rotator
Supplier: BOEKEL SCIENTIFIC