Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 | 0.01 mg, max. load 10.1 | 32 g. Draft Shield D & Activated Ionizer 1 * 1 items
Catalog # SARBMCA36P3S0024I
Supplier: Sartorius Balances
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 | 0.01 mg, max. load 10.1 | 32 g. Draft Shield D & Activated Ionizer 1 * 1 items
Catalog # SARBMCA36P3S0024I
Supplier: Sartorius Balances
Frequently Bought Together
VWR® INCU-Line® IL 10, IL 23, IL 56, Digital Incubators
Supplier: VWR Collection
Accessories for WTB Water Baths
Supplier: MEMMERT
Overhead stirrer tool, radial stirrer
Supplier: BOCHEM
Block Heater Insert, Double Wide, 384 Well Plate 1 * 1 items
Supplier: Cole Parmer
Dri-Block Aluminum Insert, 20 x 12 mm Vial, Shallow, Flat Bottom 1 * 1 items
Supplier: Cole-Parmer
Shelves for Heratherm® incubators
Supplier: Thermo Fisher Scientific
Heating circulators, HBC series
Supplier: IKA
Accessories for Reaction Stations, RS-200 and RS-250
Supplier: Cole-Parmer
Accessories for cell disruptors Sonifier®
Supplier: BRANSON
Grinding jar EasyFit for Planetary Ball Mills, tungsten carbide, 50ml 1 * 1 items
Supplier: Retsch GmbH
Boiling baths, SBB Aqua Plus series
Supplier: GRANT INSTRUMENTS
Baths and circulators, polycarbonate, CORIO™ C-BT and CD-BT series
Supplier: JULABO GmbH
Accessories for JB Academy, JB Nova, SUB Aqua Pro, SUB Aqua Plus, JB Aqua Plus and SBB Aqua Plus baths
Supplier: GRANT INSTRUMENTS
Factory Fitted Options for Binder FP Heating Chambers
Supplier: Binder
Perforated stainless steel shelf for sn 1 * 1 items
Supplier: MEMMERT