ContactUsLinkComponent
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 | 0.01 mg, max. load 10.1 | 32 g. Draft Shield D & Activated Ionizer and Draft Shield Motors 1 * 1 items
  SARBMCA36P3CF12H
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 | 0.01 mg, max. load 10.1 | 32 g. Draft Shield D & Activated Ionizer and Draft Shield Motors 1 * 1 items
  SARBMCA36P3CF12H
Frequently Bought Together