Benzyldimethyl-n-dodecylammonium chloride, 98% 1 * 1 items
Catalog # H60196.PG
Supplier: Thermo Fisher Scientific
Benzyldimethyl-n-dodecylammonium chloride, 98% 1 * 1 items
Catalog # H60196.PG
Supplier: Thermo Fisher Scientific
Frequently Bought Together
1,4-Dioxane stabilised, EMPLURA®, Supelco®
Supplier: Merck
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Chloroform analytical reagent, Supelco®
Supplier: Merck
N,N-Dimethylformamide ≥99.8% for peptide synthesis
Supplier: VWR Chemicals
Methanol, anhydrous (max. 0.003% H₂O) analytical reagent
Supplier: Merck
n-Hexane ≥99.0%, SPECTRONORM® for spectroscopy
Supplier: VWR Chemicals
Butyl acetate ≥99.5% (by GC), EMSURE® for analysis, Supelco®
Supplier: Merck
Ethanol absolute ≥99.5%, TechniSolv®, pure
Supplier: VWR Chemicals
2,2,4-Trimethylpentane, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Ethanol 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals
N,N-Dimethylacetamide, extra dry 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
Chloroform 99.8% stabilised for analysis
Supplier: Thermo Fisher Scientific
Water, TraceSELECT™ for trace analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals