ContactUsLinkComponent
Heat sealing foil for sample storage, long term storage and low temperature transportation, aluminum/adhesive lacquer, piercable 1 * 100 items
Catalog # 391-0697
Supplier:  Brand
Heat sealing foil for sample storage, long term storage and low temperature transportation, aluminum/adhesive lacquer, piercable 1 * 100 items
Catalog # 391-0697
Supplier:  Brand