The PF-2 material blend used in this Face Mask is made of rayon and laminated with an acrylic binder that has low particulate and sh edding characteristics. The Face Mask allows for excellent breathability comfort and protective while maintaining fil 1 * 500 items
Catalog # VELTPF-2SM-2-2027
Supplier: VELTEK ASSOCIATES
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.